Assay Details
Target Gene Details
Entrez Gene ID: | 212139 |
Gene Name: | coiled-coil and C2 domain containing 1A |
Gene Aliases: |
BC016188, Freud-1, Tape |
Location: |
Chr.8:84132828-84147931 on Build GRCm38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cc2d1a | NM_145970.1 | 29 | 3149 | NP_666082.2 |
| BC016188.1 | 29 | 3149 | AAH16188.1 | |
| BC027028.1 | 28 | 2964 | AAH27028.1 | |
| BC030439.1 | 11 | 1414 | AAH30439.1 | |
| DQ329239.1 | 22 | 2199 | ABC56419.1 |
Target Gene Details
Entrez Gene ID: | 244550 |
Gene Name: | podocan-like 1 |
Gene Aliases: |
5832418A03Rik, Gm506 |
Location: |
Chr.8:84125927-84143016 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Podnl1 |

Genomic Map