Assay Details
Target Gene Details
Entrez Gene ID: | 235674 |
Gene Name: | acetyl-Coenzyme A acyltransferase 1B |
Gene Aliases: |
- |
Location: |
Chr.9:119148043-119157093 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acaa1b | NM_146230.3 | 12 | 1610 | NP_666342.1 |
| AK145953.1 | 12 | 1539 | BAE26781.1 | |
| AK160568.1 | 12 | 1457 | BAE35878.1 | |
| AY273812.1 | 12 | 1608 | AAP31669.1 | |
| BC019882.1 | 12 | 1523 | AAH19882.1 |
Target Gene Details
Entrez Gene ID: | 320256 |
Gene Name: | deleted in lung and esophageal cancer 1 |
Gene Aliases: |
D630005C06Rik |
Location: |
Chr.9:119101535-119148246 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dlec1 |
More Information
Set Membership: |
|

Genomic Map