Assay Details
Target Gene Details
Entrez Gene ID: | 110446 |
Gene Name: | acetyl-Coenzyme A acetyltransferase 1 |
Gene Aliases: |
6330585C21Rik, Acat |
Location: |
Chr.9:53580522-53610350 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acat1 | NM_144784.3 | 12 | 1950 | NP_659033.1 |
| AK032097.1 | 12 | 1921 | BAC27697.1 | |
| AK075593.1 | 12 | 1923 | ||
| AK081715.1 | 12 | 1950 | BAC38304.1 | |
| AK169771.1 | 12 | 1943 | BAE41356.1 | |
| BC024763.1 | 12 | 1922 | AAH24763.1 |
More Information
Set Membership: |
|

Genomic Map