Assay Details
Target Gene Details
Entrez Gene ID: | 234733 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 19b |
Gene Aliases: |
2810457M08Rik, 4921519L13Rik, AW260119 |
Location: |
Chr.8:111003186-111031751 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx19b | NM_001190786.1 | 12 | 2307 | NP_001177715.1 |
NM_001190800.1 | 12 | 2345 | NP_001177729.1 | |
NM_172284.5 | 12 | 2350 | NP_758488.1 | |
AK033235.1 | 12 | 2308 | BAC28204.1 | |
AK147499.1 | 12 | 2345 | BAE27953.1 | |
BC025594.1 | 12 | 2310 | AAH25594.1 |
