Assay Details
Target Gene Details
Entrez Gene ID: | 234542 |
Gene Name: | retbindin |
Gene Aliases: |
A330096I21Rik, BC022744, Rtbnd |
Location: |
Chr.8:84946263-84956606 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rtbdn | NM_144929.2 | 2 | 559 | NP_659178.1 |
AK080715.1 | ||||
AY527275.1 | 2 | 559 | AAS20324.1 | |
AY527276.1 | AAS20325.1 | |||
BC022744.1 | 1 | 131 | AAH22744.1 | |
BC023051.1 | 1 | 87 | AAH23051.1 | |
BC027079.1 | 1 | 87 | AAH27079.1 |
