Assay Details
Target Gene Details
Entrez Gene ID: | 13076 |
Gene Name: | cytochrome P450, family 1, subfamily a, polypeptide 1 |
Gene Aliases: |
AHH, AHRR, CP11, Cyp1a2, P450-1 |
Location: |
Chr.9:57687928-57703824 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp1a1 | NM_001136059.2 | 7 | 1988 | NP_001129531.1 |
| NM_009992.4 | 7 | 2067 | NP_034122.1 | |
| AK005000.1 | 7 | 2068 | BAB23734.1 | |
| K02588.1 | 7 | 1987 | AAA37506.1 | |
| Y00071.1 | 7 | 1988 | CAA68277.1 |

Genomic Map