Assay Details
Target Gene Details
Entrez Gene ID: | 74032 |
Gene Name: | short chain dehydrogenase/reductase family 42E, member 1 |
Gene Aliases: |
4632417N05Rik, AI117603 |
Location: |
Chr.8:117661399-117673718 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sdr42e1 | NM_028725.3 | 3 | 809 | NP_083001.1 |
| AK014586.1 | 3 | 811 | BAB29446.1 | |
| BC038819.1 | 3 | 548 | AAH38819.1 |
More Information
Set Membership: |
|

Genomic Map