Assay Details
Target Gene Details
Entrez Gene ID: | 114255 |
Gene Name: | docking protein 4 |
Gene Aliases: |
- |
Location: |
Chr.8:94863828-94876312 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dok4 | NM_053246.3 | 9 | 1667 | NP_444476.1 |
| AF418207.1 | 8 | 1551 | AAL14626.1 | |
| AK032352.1 | 9 | 1657 | BAC27829.1 | |
| AK077612.1 | 9 | 1659 | ||
| AK139363.1 | 8 | 1556 | ||
| AK142621.1 | 8 | 1574 | BAE25134.1 | |
| AK156485.1 | 9 | 1667 | BAE33728.1 | |
| BC004705.1 | 9 | 1628 | AAH04705.1 |

Genomic Map