Assay Details
Target Gene Details
Entrez Gene ID: | 59035 |
Gene Name: | coactivator-associated arginine methyltransferase 1 |
Gene Aliases: |
Prmt4 |
Location: |
Chr.9:21546894-21589467 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Carm1 | NM_021531.6 | 16 | 2614 | NP_067506.2 |
| NM_153141.1 | 15 | 2545 | NP_694781.1 | |
| AF117887.2 | 16 | 2524 | AAD41265.2 | |
| AK158242.1 | 16 | 2579 | ||
| AK158757.1 | 16 | 2454 | BAE34644.1 | |
| BC003964.1 | 11 | 1721 | AAH03964.1 | |
| BC008263.1 | 6 | 1220 | AAH08263.1 | |
| BC036974.1 | 15 | 2529 | AAH36974.1 |
Target Gene Details
Entrez Gene ID: | 74766 |
Gene Name: | Yip1 domain family, member 2 |
Gene Aliases: |
1300010K09Rik |
Location: |
Chr.9:21588682-21592831 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Yipf2 | NM_001205157.1 | 9 | 1860 | NP_001192086.1 |
| NM_001205158.1 | 10 | 1774 | NP_001192087.1 | |
| NM_138303.2 | 8 | 1998 | NP_612176.1 | |
| NR_038062.1 | 9 | 1723 | ||
| AK004959.1 | 9 | 1708 | ||
| AK133631.1 | 9 | 1820 | BAE21759.1 | |
| BC002282.1 | 8 | 1913 | AAH02282.1 | |
| BC003289.1 | 10 | 1715 | AAH03289.1 | |
| CD774249.1 | 1 | 197 |
More Information
Set Membership: |
|

Genomic Map