Assay Details
Target Gene Details
Entrez Gene ID: | 215031 |
Gene Name: | vestigial like 2 homolog (Drosophila) |
Gene Aliases: |
C130057C21Rik, VITO-1, Vito1, vgl-2 |
Location: |
Chr.10:52022502-52028471 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vgll2 | NM_001300957.1 | 3 | 1299 | NP_001287886.1 |
| NM_153786.2 | 3 | 1302 | NP_722481.1 | |
| AF542181.1 | 3 | 1118 | AAN37898.1 | |
| AJ578054.1 | 3 | 1120 | CAE17332.1 | |
| AK048390.1 | 3 | 1274 | BAC33319.1 | |
| AK052987.1 | 3 | 1274 | BAC35232.1 | |
| BC133696.1 | 4 | 638 |
More Information
Set Membership: |
|

Genomic Map