Assay Details
Target Gene Details
Entrez Gene ID: | 213783 |
Gene Name: | pleckstrin homology domain containing, family G (with RhoGef domain) member 1 |
Gene Aliases: |
D10Ertd733e, Gm521, mKIAA1209 |
Location: |
Chr.10:3740251-3967303 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plekhg1 | NM_001033253.3 | 2 | 316 | NP_001028425.1 |
| NM_001159942.1 | 3 | 482 | NP_001153414.1 | |
| AK033676.1 | 3 | 482 | ||
| AK034403.1 | 3 | 483 | BAE20491.1 | |
| AK045140.1 | 3 | 662 | BAE20649.1 | |
| AK135923.1 | 2 | 318 | BAE22726.1 |

Genomic Map