Assay Details
Target Gene Details
Entrez Gene ID: | 235293 |
Gene Name: | sterol-C5-desaturase (fungal ERG3, delta-5-desaturase) homolog (S. cerevisae) |
Gene Aliases: |
A830037K02, A830073K23Rik, Sc5dl |
Location: |
Chr.9:42251592-42264300 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sc5d | NM_172769.2 | 6 | 1985 | NP_766357.1 |
| AK043825.1 | 6 | 1944 | BAC31666.1 | |
| AK050439.1 | ||||
| AK077670.1 | 6 | 1943 | BAC36944.1 | |
| AK154309.1 | 6 | 1932 | BAE32506.1 | |
| AK165139.1 |
More Information
Set Membership: |
|

Genomic Map