Assay Details
Target Gene Details
Entrez Gene ID: | 216021 |
Gene Name: | storkhead box 1 |
Gene Aliases: |
4732470K04Rik, Gm63 |
Location: |
Chr.10:62659043-62726099 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Stox1 | NM_001033260.1 | 1 | 170 | NP_001028432.1 |
BC137978.1 | 1 | 170 |
