Assay Details
Target Gene Details
Entrez Gene ID: | 12822 |
Gene Name: | collagen, type XVIII, alpha 1 |
Gene Aliases: |
- |
Location: |
Chr.10:77052178-77166530 on Build GRCm38 |
Assay Gene Location: | Within Exon 43 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Col18a1 | NM_001109991.1 | 41 | 5429 | NP_001103461.1 |
| NM_009929.3 | 42 | 4845 | NP_034059.2 | |
| BC064817.1 | 42 | 4816 | AAH64817.1 | |
| BC066080.1 | 42 | 4811 | AAH66080.1 | |
| D17546.1 | 40 | 4239 | BAA04483.1 |
More Information
Set Membership: |
|

Genomic Map