Assay Details
Target Gene Details
Entrez Gene ID: | 14360 |
Gene Name: | Fyn proto-oncogene |
Gene Aliases: |
AI448320, AW552119 |
Location: |
Chr.10:39369770-39565381 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fyn | NM_001122892.1 | 13 | 2103 | NP_001116364.1 |
| NM_001122893.1 | 14 | 2182 | NP_001116365.1 | |
| NM_008054.2 | 14 | 2173 | NP_032080.2 | |
| AK156584.1 | 14 | 2184 | BAE33766.1 | |
| AK171646.1 | 14 | 2175 | BAE42585.1 | |
| AK178286.1 | 1 | 59 | ||
| AK192068.1 | 1 | 71 | ||
| AK197042.1 | 1 | 71 | ||
| BC032149.1 | 13 | 1818 | AAH32149.1 | |
| BC092217.1 | 13 | 2099 | AAH92217.1 | |
| M27266.1 | 15 | 1885 | AAA37644.1 | |
| U70324.1 | 14 | 1886 | AAB09568.1 |

Genomic Map