Assay Details
Target Gene Details
Entrez Gene ID: | 14360 |
Gene Name: | Fyn proto-oncogene |
Gene Aliases: |
AI448320, AW552119 |
Location: |
Chr.10:39369770-39565381 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Fyn | NM_001122892.1 | 3 | 560 | NP_001116364.1 |
NM_001122893.1 | 4 | 630 | NP_001116365.1 | |
NM_008054.2 | 4 | 630 | NP_032080.2 | |
AK156584.1 | 4 | 632 | BAE33766.1 | |
AK171646.1 | 4 | 632 | BAE42585.1 | |
AK179168.1 | ||||
AK181277.1 | 2 | 179 | ||
AK185949.1 | ||||
AK211878.1 | 2 | 160 | ||
BC032149.1 | 3 | 275 | AAH32149.1 | |
BC092217.1 | 3 | 556 | AAH92217.1 | |
M27266.1 | 4 | 342 | AAA37644.1 | |
U70324.1 | 4 | 343 | AAB09568.1 |
