Assay Details
Target Gene Details
Entrez Gene ID: | 17123 |
Gene Name: | mucosal vascular addressin cell adhesion molecule 1 |
Gene Aliases: |
AV211525 |
Location: |
Chr.10:79664395-79668541 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Madcam1 | NM_013591.2 | 3 | 635 | NP_038619.2 |
AK137028.1 | 3 | 636 | BAE23208.1 | |
AK156096.1 | 3 | 613 | BAE33586.1 | |
BC024372.1 | 3 | 628 | AAH24372.1 | |
BC115910.1 | 3 | 629 | ||
BC118056.1 | 3 | 629 | ||
BE632565.1 | ||||
L21203.1 | 3 | 632 | AAA39485.1 | |
S80258.1 | 3 | 602 | AAB35609.1 |
