Assay Details
Target Gene Details
Entrez Gene ID: | 17123 |
Gene Name: | mucosal vascular addressin cell adhesion molecule 1 |
Gene Aliases: |
AV211525 |
Location: |
Chr.10:79664395-79668541 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Madcam1 | NM_013591.2 | 5 | 1173 | NP_038619.2 |
| AK137028.1 | 5 | 1174 | BAE23208.1 | |
| AK156096.1 | 4 | 719 | BAE33586.1 | |
| BC024372.1 | 4 | 734 | AAH24372.1 | |
| BC115910.1 | 4 | 735 | ||
| BC118056.1 | 4 | 735 | ||
| L21203.1 | 5 | 1170 | AAA39485.1 | |
| S80258.1 | 4 | 708 | AAB35609.1 |

Genomic Map