Assay Details
Target Gene Details
Entrez Gene ID: | 16173 |
Gene Name: | interleukin 18 |
Gene Aliases: |
Igif, Il-18 |
Location: |
Chr.9:50554705-50581841 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il18 | AK078100.1 |
Target Gene Details
Entrez Gene ID: | 66654 |
Gene Name: | testis expressed gene 12 |
Gene Aliases: |
5730518K06Rik |
Location: |
Chr.9:50557148-50561268 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tex12 | NM_025687.3 | 5 | 388 | NP_079963.1 |
| AF285582.1 | 5 | 446 | AAK31961.1 | |
| AK006285.1 | 5 | 432 | BAB24504.1 | |
| AK014896.1 | 5 | 445 | BAB29611.1 | |
| AK017776.1 | 5 | 444 | BAB30924.1 | |
| BC048458.1 | 5 | 388 | AAH48458.1 | |
| BC061081.1 | 5 | 443 | AAH61081.1 |

Genomic Map