Assay Details
Target Gene Details
Entrez Gene ID: | 12905 |
Gene Name: | CASP2 and RIPK1 domain containing adaptor with death domain |
Gene Aliases: |
RAIDD |
Location: |
Chr.10:95174739-95324345 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cradd | NM_009950.2 | 3 | 832 | NP_034080.1 |
AJ224738.1 | 3 | 831 | CAA12107.1 | |
AJ224739.1 | 2 | 568 | CAA12108.1 | |
AJ224740.1 | 3 | 817 | CAA12109.1 | |
AK136561.1 | 3 | 737 | BAE23045.1 | |
AK163112.1 | 3 | 844 | BAE37198.1 | |
BC005608.1 | 2 | 795 | AAH05608.1 |
