Assay Details
Target Gene Details
Entrez Gene ID: | 23887 |
Gene Name: | gamma-glutamyltransferase 5 |
Gene Aliases: |
AI551243, GGL, GGT-REL, Ggtla1 |
Location: |
Chr.10:75589327-75616968 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ggt5 | NM_011820.4 | 12 | 2268 | NP_035950.2 |
AF077765.1 | 12 | 2253 | AAC71001.1 | |
AK029138.1 | 5 | 804 | BAC26318.1 | |
AK049084.1 | 12 | 2272 | ||
AK052183.1 | 12 | 2269 | BAC34873.1 | |
AK140308.1 | 11 | 2789 | ||
AK170326.1 | 12 | 2132 | BAE41720.1 | |
BC051971.1 | 11 | 1963 | AAH51971.1 | |
BC113775.1 | 12 | 1992 |
