Assay Details
Target Gene Details
Entrez Gene ID: | 319468 |
Gene Name: | protein phosphatase 1H (PP2C domain containing) |
Gene Aliases: |
A430075L18Rik, ARHCL1, C030002B11Rik |
Location: |
Chr.10:122678106-122945795 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppm1h | NM_001110218.1 | 10 | 5629 | NP_001103688.1 |
| AK088561.1 | 1 | 365 |
More Information
Set Membership: |
|

Genomic Map