Assay Details
Target Gene Details
Entrez Gene ID: | 319468 |
Gene Name: | protein phosphatase 1H (PP2C domain containing) |
Gene Aliases: |
A430075L18Rik, ARHCL1, C030002B11Rik |
Location: |
Chr.10:122678106-122945795 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppm1h | NM_001110218.1 | 1 | 448 | NP_001103688.1 |
| NM_176919.4 | 1 | 448 | NP_795893.2 | |
| AK029461.1 | BAE43270.1 | |||
| AK040194.1 | 1 | 192 | BAC30536.1 | |
| AK040207.1 | 1 | 433 | BAE43308.1 | |
| AK043121.1 | 1 | 449 | ||
| AK051434.1 | 1 | 406 | ||
| AK077422.1 | 1 | 433 | ||
| AK134804.1 | 1 | 428 | BAE22292.1 | |
| AK150309.1 | BAE29457.1 | |||
| AK220260.1 | 1 | 405 | BAD90185.1 |
More Information
Set Membership: |
|

Genomic Map