Assay Details
Target Gene Details
Entrez Gene ID: | 67557 |
Gene Name: | La ribonucleoprotein domain family, member 6 |
Gene Aliases: |
5430431G03Rik, AI552438, Achn |
Location: |
Chr.9:60713121-60738801 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Larp6 | NM_026235.4 | 3 | 1398 | NP_080511.2 |
| AK017372.1 | 3 | 1403 | BAB30713.1 | |
| AK039614.1 | 3 | 1381 | BAC30400.1 | |
| AK042616.1 | 3 | 1330 | BAC31306.1 | |
| AK047452.1 | 1 | 702 | BAC33063.1 | |
| AK087521.1 | 3 | 1401 | BAC39909.1 | |
| BC090615.1 | 3 | 1530 | AAH90615.1 |
More Information
Set Membership: |
|

Genomic Map