Assay Details
Target Gene Details
Entrez Gene ID: | 66268 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class Y-like |
Gene Aliases: |
1810008A14Rik, PIG-Y, Pigy |
Location: |
Chr.9:22156846-22158355 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pigyl | NM_001082532.1 | 2 | 594 | NP_001076001.1 |
| AI414555.1 | 1 | 99 | ||
| AK007367.1 | 2 | 575 | BAB24989.1 | |
| AK021110.1 | 2 | 575 | BAB32300.1 | |
| BU558514.1 | 2 | 597 |
More Information
Set Membership: |
|

Genomic Map