Assay Details
Target Gene Details
Entrez Gene ID: | 64009 |
Gene Name: | spectrin repeat containing, nuclear envelope 1 |
Gene Aliases: |
8B, A330049M09Rik, BE692247, C130039F11Rik, CPG2, Myne1, mKIAA1756 |
Location: |
Chr.10:5020192-5550829 on Build GRCm38 |
Assay Gene Location: | Within Exon 88 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syne1 | NM_001079686.1 | 55 | 10529 | NP_001073154.1 |
| NM_022027.2 | 17 | 4483 | NP_071310.2 | |
| AK082196.1 | 1 | 437 |

Genomic Map