Assay Details
Target Gene Details
Entrez Gene ID: | 103236 |
Gene Name: | casein kinase 1, gamma 2 |
Gene Aliases: |
2810429I12Rik, AI463719 |
Location: |
Chr.10:80622705-80640771 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Csnk1g2 | NM_001159591.1 | 12 | 2130 | NP_001153063.1 |
| NM_134002.2 | 11 | 2307 | NP_598763.1 | |
| AK013192.1 | 1 | 280 | ||
| AK152413.1 | 11 | 2306 | ||
| BC004839.1 | 11 | 2249 | AAH04839.1 | |
| BC005750.1 | 9 | 1336 | AAH05750.1 | |
| CF612914.1 | 1 | 250 |
More Information
Set Membership: |
|

Genomic Map