Assay Details
Target Gene Details
Entrez Gene ID: | 83554 |
Gene Name: | follistatin-like 3 |
Gene Aliases: |
E030038F23Rik, Flrg |
Location: |
Chr.10:79777274-79782630 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fstl3 | NM_031380.2 | 5 | 1125 | NP_113557.1 |
| AF276238.1 | 5 | 1104 | AAG47666.1 | |
| AK079286.1 | 3 | 745 | ||
| AK082898.1 | 5 | 1127 | BAC38676.1 | |
| AK087247.1 | 5 | 1109 | BAC39827.1 | |
| BC061052.1 | 5 | 1125 | AAH61052.1 |
Target Gene Details
Entrez Gene ID: | 73106 |
Gene Name: | protease, serine 57 |
Gene Aliases: |
2900092M14Rik, GLGL782, Prssl1, UNQ782 |
Location: |
Chr.10:79781474-79795605 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prss57 | NM_001042710.1 | 5 | 2365 | NP_001036175.1 |
| AK158376.1 | 5 | 2366 |
More Information
Set Membership: |
|

Genomic Map