Assay Details
Target Gene Details
Entrez Gene ID: | 100038468 |
Gene Name: | predicted gene 10684 |
Gene Aliases: |
ENSMUSG00000074385 |
Location: |
Chr.9:45107176-45135606 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm10684 | NR_033547.1 | |||
| AK039498.1 | ||||
| AK045437.1 | BAE20650.1 |
Target Gene Details
Entrez Gene ID: | 72821 |
Gene Name: | sodium channel, voltage-gated, type II, beta |
Gene Aliases: |
2810451E09Rik, AI840361, Gm183 |
Location: |
Chr.9:45117876-45130070 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Scn2b | NM_001014761.2 | 4 | 2576 | NP_001014761.1 |
More Information
Set Membership: |
|

Genomic Map