Assay Details
Target Gene Details
Entrez Gene ID: | 70097 |
Gene Name: | SAM and SH3 domain containing 1 |
Gene Aliases: |
1100001C18Rik, 2500002E12Rik, A330076K04Rik, mKIAA0790 |
Location: |
Chr.10:8722217-8956137 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sash1 | NM_175155.4 | 1 | 60 | NP_780364.3 |
AK010852.1 | 2 | 623 | BAC25318.1 | |
AK049453.1 | ||||
AK149872.1 | ||||
CJ125950.1 | 1 | 61 |
