Assay Details
Target Gene Details
Entrez Gene ID: | 215821 |
Gene Name: | ARFGEF family member 3 |
Gene Aliases: |
9630004D11, B930094H20Rik, BIG3, D10Bwg1379e, mKIAA1244 |
Location: |
Chr.10:18581839-18744069 on Build GRCm38 |
Assay Gene Location: | Within Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arfgef3 | NM_001033258.4 | 34 | 7340 | NP_001028430.1 |
| AK035782.1 | 10 | 2039 | ||
| AK122475.1 | 23 | 5702 | BAC65757.1 | |
| AK147679.1 | 34 | 7341 | BAE28069.1 | |
| AK165602.1 | 33 | 6069 |
More Information
Set Membership: |
|

Genomic Map