Assay Details
Target Gene Details
Entrez Gene ID: | 15182 |
Gene Name: | histone deacetylase 2 |
Gene Aliases: |
D10Wsu179e, YAF1, Yy1bp, mRPD3 |
Location: |
Chr.10:36974544-37001889 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac2 | NM_008229.2 | 1 | 220 | NP_032255.2 |
AB221663.1 | ||||
AK051753.1 | 1 | 195 | BAC34755.1 | |
AK165734.1 | 1 | 221 | BAE38360.1 | |
BC138517.1 | 1 | 192 | ||
U31758.1 | 1 | 187 | AAC52889.1 |
