Assay Details
Target Gene Details
Entrez Gene ID: | 15182 |
Gene Name: | histone deacetylase 2 |
Gene Aliases: |
D10Wsu179e, YAF1, Yy1bp, mRPD3 |
Location: |
Chr.10:36974544-37001889 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hdac2 | NM_008229.2 | 14 | 1706 | NP_032255.2 |
AB221663.1 | ||||
AK141649.1 | 4 | 323 | BAE24786.1 | |
BC138517.1 | 14 | 1678 | ||
U31758.1 | 14 | 1666 | AAC52889.1 |
