Assay Details
Target Gene Details
Entrez Gene ID: | 70097 |
Gene Name: | SAM and SH3 domain containing 1 |
Gene Aliases: |
1100001C18Rik, 2500002E12Rik, A330076K04Rik, mKIAA0790 |
Location: |
Chr.10:8722217-8956137 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sash1 | NM_175155.4 | 18 | 2589 | NP_780364.3 |
AJ507736.1 | 18 | 2404 | CAD47812.1 | |
AK039638.1 | 6 | 928 | ||
AK129216.1 | 13 | 1923 | BAC98026.1 | |
AK149872.1 | 18 | 2541 |
