Assay Details
Target Gene Details
Entrez Gene ID: | 116848 |
Gene Name: | bromodomain adjacent to zinc finger domain, 2A |
Gene Aliases: |
AA415431, C030005G16Rik, C78388, Tip5, Walp3, mKIAA0314 |
Location: |
Chr.10:128091056-128129304 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Baz2a | NM_054078.2 | 25 | 4808 | NP_473419.2 |
| AJ309544.2 | 23 | 4542 | CAC69992.1 | |
| AK084735.1 | ||||
| AK122243.1 | 8 | 1654 | BAC65525.1 | |
| AK135732.1 | 5 | 818 | BAE22634.1 | |
| AK140558.1 | 4 | 625 | BAE24422.1 | |
| AK149353.1 | 2 | 182 | BAE28829.1 |
More Information
Set Membership: |
|

Genomic Map