Assay Details
Target Gene Details
Entrez Gene ID: | 15273 |
Gene Name: | human immunodeficiency virus type I enhancer binding protein 2 |
Gene Aliases: |
MIBP1, Schnurri-2, Shn-2 |
Location: |
Chr.10:13965779-14151378 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Hivep2 | NM_010437.2 | 4 | 5049 | NP_034567.2 |
AK147244.1 | 5 | 5130 | BAE27792.1 | |
AK147348.1 | 4 | 5049 | ||
AK147419.1 | 9 | 5339 | BAE27900.1 | |
AK147655.1 | 4 | 5049 | ||
BC150704.1 | 1 | 4254 | ||
Y15907.1 | 1 | 4300 | CAA75868.1 |
