Assay Details
Target Gene Details
Entrez Gene ID: | 71664 |
Gene Name: | methyltransferase like 7B |
Gene Aliases: |
0610006F02Rik, AI266817 |
Location: |
Chr.10:128958277-128960988 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mettl7b | NM_027853.2 | 1 | 183 | NP_082129.2 |
| AK002239.1 | 1 | 184 | BAB21956.2 | |
| BC024898.1 | 1 | 68 | AAH24898.1 |
More Information
Set Membership: |
|

Genomic Map