Assay Details
Target Gene Details
Entrez Gene ID: | 21780 |
Gene Name: | transcription factor A, mitochondrial |
Gene Aliases: |
AI661103, Hmgts, mtTFA, tsHMG |
Location: |
Chr.10:71225468-71238957 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Tfam | NM_009360.4 | 7 | 2850 | NP_033386.1 |
AK032682.1 | 2 | 2189 | ||
AK050446.1 | 7 | 2851 | BAC34258.1 |
