Assay Details
Target Gene Details
Entrez Gene ID: | 76964 |
Gene Name: | RIKEN cDNA 2610028H24 gene |
Gene Aliases: |
ORF67 |
Location: |
Chr.10:76449023-76461218 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2610028H24Rik | NM_029816.2 | 8 | 1395 | NP_084092.2 |
| AK011591.1 | 8 | 1348 | BAB27720.1 | |
| AK161552.1 | 8 | 1205 | BAE36458.1 | |
| BC051454.1 | 8 | 1329 | AAH51454.1 | |
| BQ552827.1 | 1 | 369 |
Target Gene Details
Entrez Gene ID: | 216119 |
Gene Name: | ybeY metallopeptidase |
Gene Aliases: |
1110068N23Rik, A130042E20Rik, C21orf57, ORF57, ORF66 |
Location: |
Chr.10:76459567-76469114 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ybey | NM_172550.4 | 5 | 3239 | NP_766138.3 |
| AK037727.1 | 5 | 2966 | BAC29859.1 | |
| AK149490.1 | 1 | 745 | ||
| BC057308.1 | 5 | 3239 | AAH57308.1 |
More Information
Set Membership: |
|

Genomic Map