Assay Details
Target Gene Details
Entrez Gene ID: | 237400 |
Gene Name: | mex3 homolog D (C. elegans) |
Gene Aliases: |
BC059858, Rkhd1 |
Location: |
Chr.10:80380355-80387651 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mex3d | NM_198615.2 | 1 | 826 | NP_941017.2 |
| AK149801.1 | 1 | 354 | BAE29094.1 |
More Information
Set Membership: |
|

Genomic Map