Assay Details
Target Gene Details
Entrez Gene ID: | 30055 |
Gene Name: | translocase of inner mitochondrial membrane 13 |
Gene Aliases: |
D10Ertd378e, Tim9, Timm9 |
Location: |
Chr.10:80899450-80900969 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Timm13 | NM_013895.4 | 3 | 555 | NP_038923.1 |
AF144702.1 | 3 | 557 | AAD39953.1 | |
AK003054.1 | 3 | 361 | BAB22536.1 | |
AK009041.1 | 3 | 349 | BAB26042.1 | |
AK009152.1 | 3 | 365 | BAB26109.1 | |
AK013230.1 | 3 | 353 | BAB28728.1 | |
AK088984.1 | 3 | 365 | BAC40687.1 | |
AK213641.1 | ||||
BC011436.1 | 3 | 368 | AAH11436.1 |
