Assay Details
Target Gene Details
Entrez Gene ID: | 19279 |
Gene Name: | protein tyrosine phosphatase, receptor type, R |
Gene Aliases: |
Gmcp1, PTP-SL, PTPBR7, RPTPRR, mPTP213 |
Location: |
Chr.10:116018200-116274932 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptprr | NM_011217.2 | 1 | 375 | NP_035347.1 |
| AK028054.1 | 1 | 376 | BAC25724.1 | |
| AK051719.1 | 1 | 452 | ||
| BC119230.1 | 1 | 308 | ||
| BC120547.1 | 1 | 308 | ||
| D31898.1 | 1 | 338 | BAA06696.1 |

Genomic Map