Assay Details
Target Gene Details
Entrez Gene ID: | 237353 |
Gene Name: | SH3 domain containing ring finger 3 |
Gene Aliases: |
4831416G18Rik, Igfbp5-ip, Sh3md4, Sh3rf31 |
Location: |
Chr.10:58812372-59138916 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sh3rf3 | NM_172788.3 | 8 | 2183 | NP_766376.2 |
AK029202.1 | 8 | 2086 | BAC26346.1 | |
AK132983.1 | 6 | 1190 | BAE21454.1 | |
BC057304.1 | 8 | 1439 | AAH57304.1 |
