Assay Details
Target Gene Details
Entrez Gene ID: | 100503659 |
Gene Name: | downstream of Stk11 |
Gene Aliases: |
9630008F14, R29144/1 |
Location: |
Chr.10:80130434-80140136 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dos | NM_001195268.1 | 9 | 1750 | NP_001182197.1 |
| NR_036582.1 | 11 | 2086 | ||
| AK035824.1 | 3 | 901 | BAC29200.1 | |
| AK137009.1 | 11 | 2087 | ||
| BC078442.1 | 5 | 1305 | AAH78442.1 | |
| BC100552.1 | 4 | 1133 | AAI00553.1 |
More Information
Set Membership: |
|

Genomic Map