Assay Details
Target Gene Details
Entrez Gene ID: | 208266 |
Gene Name: | DOT1-like, histone H3 methyltransferase (S. cerevisiae) |
Gene Aliases: |
A630076O07, AW907654, Dot1, KMT4, mDot1 |
Location: |
Chr.10:80755129-80795461 on Build GRCm38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dot1l | NM_199322.1 | 20 | 2555 | NP_955354.1 |
| AK042267.1 | 8 | 1323 | ||
| AK220271.1 | 20 | 2513 | BAD90196.1 | |
| AY196089.1 | 20 | 2555 | AAP42293.1 | |
| AY376663.1 | 17 | 2372 | AAQ84680.1 | |
| AY377920.1 | 13 | 1677 | AAQ82850.1 | |
| JF508521.1 | 20 | 2317 |
More Information
Set Membership: |
|

Genomic Map