Assay Details
Target Gene Details
Entrez Gene ID: | 320183 |
Gene Name: | methionine sulfoxide reductase B3 |
Gene Aliases: |
D430026P16Rik, I-8-23, MsrB2 |
Location: |
Chr.10:120781098-120899304 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Msrb3 |
