Assay Details
Target Gene Details
Entrez Gene ID: | 67074 |
Gene Name: | MON2 homolog (yeast) |
Gene Aliases: |
2610528O22Rik, AW495628, Sf21, mKIAA1040 |
Location: |
Chr.10:122992060-123076505 on Build GRCm38 |
Assay Gene Location: | Within Exon 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mon2 | NM_001163024.1 | 36 | 8040 | NP_001156496.1 |
| NM_001163025.1 | 34 | 8022 | NP_001156497.1 | |
| NM_153395.2 | 34 | 8019 | NP_700444.2 |
More Information
Set Membership: |
|

Genomic Map