Assay Details
Target Gene Details
Entrez Gene ID: | 14628 |
Gene Name: | osteopetrosis associated transmembrane protein 1 |
Gene Aliases: |
1200002H13Rik, HSPC019, gl |
Location: |
Chr.10:42678916-42702462 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ostm1 | NM_172416.3 | 6 | 2336 | NP_766004.1 |
| AF533890.1 | 6 | 2335 | AAO72748.1 | |
| AK004546.1 | 6 | 2313 | BAC25087.1 | |
| AK044897.1 | 6 | 2335 | ||
| AK050513.1 | 6 | 2310 | BAC34300.1 | |
| BC057635.1 | 6 | 2274 | AAH57635.1 |

Genomic Map