Assay Details
Target Gene Details
Entrez Gene ID: | 19279 |
Gene Name: | protein tyrosine phosphatase, receptor type, R |
Gene Aliases: |
Gmcp1, PTP-SL, PTPBR7, RPTPRR, mPTP213 |
Location: |
Chr.10:116018200-116274932 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ptprr | NM_011217.2 | 2 | 646 | NP_035347.1 |
AK028054.1 | 2 | 647 | BAC25724.1 | |
AK051719.1 | 2 | 723 | ||
BC119230.1 | 2 | 579 | ||
BC120547.1 | 2 | 579 | ||
D31898.1 | 2 | 609 | BAA06696.1 |
