Assay Details
Target Gene Details
Entrez Gene ID: | 20431 |
Gene Name: | premelanosome protein |
Gene Aliases: |
D10H12S53E, D12S53Eh, Pmel17, Si, Silv, gp100, gp87 |
Location: |
Chr.10:128705168-128720238 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pmel | NM_021882.4 | 11 | 1838 | NP_068682.2 |
| AF119092.1 | 6 | 715 | AAF19761.1 | |
| AK012808.1 | 11 | 1839 | BAB28486.1 | |
| AK131956.1 | 11 | 1840 | BAE20900.1 | |
| AK181412.1 | ||||
| AK182074.1 | ||||
| AK182415.1 | ||||
| AK186888.1 | ||||
| AK191933.1 | ||||
| AK195401.1 | ||||
| AK207055.1 | ||||
| AK207213.1 | ||||
| AK208589.1 | ||||
| AK210548.1 | ||||
| AK211442.1 | ||||
| AK214408.1 | ||||
| AK218557.1 | ||||
| AK219470.1 | ||||
| AK219862.1 | ||||
| BC082555.1 | 11 | 1824 | AAH82555.1 | |
| U14133.1 | 11 | 1812 | AAA69538.1 |
More Information
Set Membership: |
|

Genomic Map